missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-10-Camphorsulfonic Acid 98.0+%, TCI America™

Catalog No. C001625G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C001625G 25 g
C0016100G 100 g
C0016500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C001625G Supplier TCI America Supplier No. C001625G

Chemical Identifiers

CAS 8-2-5872
Molecular Formula C10H16O4S
Molecular Weight (g/mol) 232.29
MDL Number MFCD00074827
InChI Key MIOPJNTWMNEORI-UHFFFAOYNA-N
Synonym reychler's acid, camphorsulfonic acid, d-camphorsulfonic acid, camphersulfosaeure, --10-camphorsulfonic acid, 2-oxobornane-10-sulphonic acid, d-10-camphorsulfonic acid, 7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonic acid, l-camphor-10-sulfonic acid, +-10-camphorsulfonic acid
PubChem CID 18462
ChEBI CHEBI:55379
IUPAC Name (7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl)methanesulfonic acid
SMILES CC1(C2CCC1(C(=O)C2)CS(=O)(=O)O)C

Specifications

Melting Point 202°C
Color White-Yellow
Quantity 25 g
UN Number 3261
Formula Weight 232.29
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (+/-)-10-Camphorsulfonic Acid
EINECSNumber (4)-0309
TSCA Yes