missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-10-Camphorsulfonic Acid 98.0+%, TCI America™

Supplier:  TCI America C0016100G

Quantity 100 g

 View more versions of this product

Catalog No. C0016100G


Only null left

Description

Description

Specifications

Chemical Identifiers

8-2-5872
232.29
MIOPJNTWMNEORI-UHFFFAOYNA-N
18462
(7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl)methanesulfonic acid
C10H16O4S
MFCD00074827
reychler's acid, camphorsulfonic acid, d-camphorsulfonic acid, camphersulfosaeure, --10-camphorsulfonic acid, 2-oxobornane-10-sulphonic acid, d-10-camphorsulfonic acid, 7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonic acid, l-camphor-10-sulfonic acid, +-10-camphorsulfonic acid
CHEBI:55379
CC1(C2CCC1(C(=O)C2)CS(=O)(=O)O)C

Specifications

(+/-)-10-Camphorsulfonic Acid
202°C
C10H16O4S
MFCD00074827
reychler's acid, camphorsulfonic acid, d-camphorsulfonic acid, camphersulfosaeure, --10-camphorsulfonic acid, 2-oxobornane-10-sulphonic acid, d-10-camphorsulfonic acid, 7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonic acid, l-camphor-10-sulfonic acid, +-10-camphorsulfonic acid
CC1(C2CCC1(C(=O)C2)CS(=O)(=O)O)C
232.29
CHEBI:55379
≥98.0% (T)
8-2-5872
White-Yellow
100 g
3261
MIOPJNTWMNEORI-UHFFFAOYNA-N
(7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl)methanesulfonic acid
18462
232.29
Crystalline Powder
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (4)-0309

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions