missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Positive Displacement Pipette Tips
Description
- Replacement tips for for 10 to 100 μL postive displacement pipettor, catalog number 07WS1075 pipettor
- Capillaries and pistons are pre-assembled and presterilized
- Come in racks of 60
Specifications
Specifications
| Capacity | 10 to 100 μL |
| For Use With (Equipment) | SafTest System |
| For Use With (Application) | Lipid degradation and oxidation |
| Packaging Type | Box |
| Product Type | Positive displacement pipet tips |
| Quantity | 60 Pk. |