missing translation for 'onlineSavingsMsg'
Learn More

1-tert-Butyl-4-nitrobenzene 97.0+%, TCI America™

Catalog No. B21495G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21495G 5 g
B214925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B21495G Supplier TCI America Supplier No. B21495G

Chemical Identifiers

CAS 3282-56-2
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.219
MDL Number MFCD00041253
InChI Key XSCPVQNNFLHGHY-UHFFFAOYSA-N
Synonym 1-tert-butyl-4-nitrobenzene, 4-t-butylnitrobenzene, 4-tert-butylnitrobenzene, p-t-butylnitrobenzene, p-nitro-t-butylbenzene, 4-t-butyl nitrobenzene, benzene, 1-1,1-dimethylethyl-4-nitro, tert-butyl-4-nitrobenzene, acmc-209hw6, benzene,1-dimethylethyl-4-nitro
PubChem CID 76773
IUPAC Name 1-tert-butyl-4-nitrobenzene
SMILES CC(C)(C)C1=CC=C(C=C1)[N+](=O)[O-]

Specifications

Color Yellow
Boiling Point 267°C
Quantity 5 g
Formula Weight 179.22
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-tert-Butyl-4-nitrobenzene