accessibility menu, dialog, popup

UserName

CAS RN 3282-56-2

IUPAC Name: 1-tert-butyl-4-nitrobenzene
Synonyms: 4-T-butylnitrobenzene
Molecular Weight (g/mol): 179.22
Molecular Formula: C10H13NO2
InChi Key: XSCPVQNNFLHGHY-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results