missing translation for 'onlineSavingsMsg'
Learn More

1-tert-Butyl-4-iodobenzene 95.0+%, TCI America™

Catalog No. B36245G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36245G 5 g
B362425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B36245G Supplier TCI America Supplier No. B36245G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 35779-04-5
Molecular Formula C10H13I
Molecular Weight (g/mol) 260.118
MDL Number MFCD00052339
InChI Key WQVIVQDHNKQWTM-UHFFFAOYSA-N
Synonym 1-tert-butyl-4-iodobenzene, 4-tert-butyliodobenzene, 4-t-butyliodobenzene, 1-iodo-4-tert-butylbenzene, benzene, 1-1,1-dimethylethyl-4-iodo, 4-tert-butyl-1-iodobenzene, 2-4-iodophenyl-2-methylpropane, 2-4'-iodophenyl-2-methylpropane, pubchem3974, 4-t-butyl-iodobenzene
PubChem CID 142029
IUPAC Name 1-tert-butyl-4-iodobenzene
SMILES CC(C)(C)C1=CC=C(C=C1)I

Specifications

Color Yellow
Boiling Point 118°C
Quantity 5 g
Formula Weight 260.12
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-tert-Butyl-4-iodobenzene