accessibility menu, dialog, popup

UserName

CH3H3CCH3I

CAS RN 35779-04-5

IUPAC Name: 1-tert-butyl-4-iodobenzene
Synonyms: 1-tert-butyl-4-iodobenzene
Molecular Weight (g/mol): 260.12
Molecular Formula: C10H13I
InChi Key: WQVIVQDHNKQWTM-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(I)C=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)