missing translation for 'onlineSavingsMsg'
Learn More

1-tert-Butyl-4-iodobenzene, 97%

Catalog No. p-7045688
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1160303 1 g
AAL1160306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1160303 Supplier Thermo Scientific Chemicals Supplier No. L1160303
Only null left

CAS: 35779-04-5 | C10H13I | 260.118 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 35779-04-5
Molecular Formula C10H13I
Molecular Weight (g/mol) 260.118
MDL Number MFCD00052339
InChI Key WQVIVQDHNKQWTM-UHFFFAOYSA-N
Synonym 1-tert-butyl-4-iodobenzene, 4-tert-butyliodobenzene, 4-t-butyliodobenzene, 1-iodo-4-tert-butylbenzene, benzene, 1-1,1-dimethylethyl-4-iodo, 4-tert-butyl-1-iodobenzene, 2-4-iodophenyl-2-methylpropane, 2-4'-iodophenyl-2-methylpropane, pubchem3974, 4-t-butyl-iodobenzene
PubChem CID 142029
IUPAC Name 1-tert-butyl-4-iodobenzene
SMILES CC(C)(C)C1=CC=C(C=C1)I

Specifications

Density 1.468
Boiling Point 243°C
Flash Point 106°C (223°F)
Refractive Index 1.571
Quantity 1 g
Beilstein 1931810
Sensitivity Light Sensitive
Formula Weight 260.12
Percent Purity 97%
Chemical Name or Material 1-tert-Butyl-4-iodobenzene
Hazard Category H319
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P264b-P280i-P305+P351+P338
TSCA No

RUO – Research Use Only