missing translation for 'onlineSavingsMsg'
Learn More

1-tert-Butyl-4-chlorobenzene 98.0+%, TCI America™

Catalog No. p-7119830
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28965G 5 g
B289625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B28965G Supplier TCI America Supplier No. B28965G

Chemical Identifiers

CAS 3972-56-3
Molecular Formula C10H13Cl
Molecular Weight (g/mol) 168.664
InChI Key XRTANKYQJQXSFP-UHFFFAOYSA-N
PubChem CID 77594
IUPAC Name 1-tert-butyl-4-chlorobenzene
SMILES CC(C)(C)C1=CC=C(C=C1)Cl

Specifications

Color White-Yellow
Boiling Point 216°C
Quantity 5 g
Formula Weight 168.66
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 1-tert-Butyl-4-chlorobenzene