accessibility menu, dialog, popup

UserName

CH3H3CCH3Cl

CAS RN 3972-56-3

IUPAC Name: 1-tert-butyl-4-chlorobenzene
Synonyms: 1-tert-butyl-4-chlorobenzene
Molecular Weight (g/mol): 168.66
Molecular Formula: C10H13Cl
InChi Key: XRTANKYQJQXSFP-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(Cl)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results