Learn More
Invitrogen™ eBioscience™ 1-step Fix/Lyse Solution (10X)
Enables lysis of RBCs after staining peripheral blood cells with fluorochrome-conjugated antibodies. Formulated to lyse non-nucleated erythrocytes while maintaining fixed and labeled leukocyte population.
$242.00 - $740.00
Specifications
Type 1-step Fixation/Lyse Solution
Product Line eBioscience
View More Specs
Alert:
Contains 10% formaldehyde and 30% diethylene glycol
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
5018784
View Documents Promotion Details
Invitrogen™
00533354
50 mL
Each for $242.00
Only null left
501128856
View Documents Promotion Details
Invitrogen™
00533357
200 mL
Each for $740.00
Only null left
Description
Description
  • Isolation of mononuclear cells by gradient centrifugation (e.g. Ficoll-Paque density gradient separation) is not required when using this solution
  • Compatible with antibodies conjugated to organic dyes and tandem dyes
Specifications
Specifications
1-step Fixation/Lyse Solution
eBioscience
Videos
SDS
Documents
Documents
For Research Use Only. Not for use in diagnostic procedures.