missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ 1-step Fix/Lyse Solution (10X)

Catalog No. 501128856
Change view
Click to view available options
Quantity:
50 mL
200 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501128856 200 mL
5018784 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Contains 10% formaldehyde and 30% diethylene glycol
Catalog No. 501128856 Supplier Invitrogen™ Supplier No. 00533357
Contains 10% formaldehyde and 30% diethylene glycol

Enables lysis of RBCs after staining peripheral blood cells with fluorochrome-conjugated antibodies. Formulated to lyse non-nucleated erythrocytes while maintaining fixed and labeled leukocyte population.

  • Isolation of mononuclear cells by gradient centrifugation (e.g. Ficoll-Paque density gradient separation) is not required when using this solution
  • Compatible with antibodies conjugated to organic dyes and tandem dyes

Specifications

Type 1-step Fixation/Lyse Solution
Product Line eBioscience
Quantity 200 mL

For Research Use Only. Not for use in diagnostic procedures.