Learn More
(1-Propyl)triphenylphosphonium bromide, 99%
CAS: 6228-47-3 | C21H22BrP | 385.29 g/mol
Supplier: Thermo Scientific Chemicals A129680B
Description
It is employed as a catalyst for the carboxylation of styrene oxide and as a reactant involved in epoxidation of arylalkenes, boration of allylic epoxides, Wittig reactions, selective dimerization of α-olefins, and synthesis of indoles. It has also been shown to be part of Phosphonium salts, having antiviral activity against influenza virus A.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is employed as a catalyst for the carboxylation of styrene oxide and as a reactant involved in epoxidation of arylalkenes, boration of allylic epoxides, Wittig reactions, selective dimerization of α-olefins, and synthesis of indoles. It has also been shown to be part of Phosphonium salts, having antiviral activity against influenza virus A.
Solubility
Soluble in water.
Notes
Hygroscopic. Store away from oxidizing agents, humidity and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Specifications
| (1-Propyl)triphenylphosphonium bromide | |
| 236°C to 240°C | |
| MFCD00011843 | |
| 3599726 | |
| propyltriphenylphosphonium bromide, triphenyl propyl phosphonium bromide, 1-propyl triphenylphosphonium bromide, n-propyltriphenylphosphonium bromide, triphenyl propyl phosphanium bromide, n-propyl triphenylphosphonium bromide, propyl triphenyl phosphonium chloride, acmc-1bff2, propyltriphenylphosphoniumbromide | |
| XMQSELBBYSAURN-UHFFFAOYSA-M | |
| triphenyl(propyl)phosphanium;bromide | |
| 80374 | |
| 99% |
| 6228-47-3 | |
| C21H22BrP | |
| 1000 g | |
| Hygroscopic | |
| Soluble in water. | |
| [Br-].CCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 385.29 | |
| 385.29 |
Chemical Identifiers
| 6228-47-3 | |
| 385.29 | |
| XMQSELBBYSAURN-UHFFFAOYSA-M | |
| 80374 | |
| [Br-].CCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
| C21H22BrP | |
| MFCD00011843 | |
| propyltriphenylphosphonium bromide, triphenyl propyl phosphonium bromide, 1-propyl triphenylphosphonium bromide, n-propyltriphenylphosphonium bromide, triphenyl propyl phosphanium bromide, n-propyl triphenylphosphonium bromide, propyl triphenyl phosphonium chloride, acmc-1bff2, propyltriphenylphosphoniumbromide | |
| triphenyl(propyl)phosphanium;bromide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 228-330-2
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only