missing translation for 'onlineSavingsMsg'
Learn More

1-Propenylmagnesium bromide, 0.5M solution in THF, AcroSeal™

Catalog No. AC434678000
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC434678000 800 mL AcroSeal™ Glass Bottle
AC434671000 100 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC434678000 Supplier Thermo Scientific Chemicals Supplier No. 434678000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 14092-04-7 | C3H5BrMg | 145.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14092-04-7 , 109-99-9
Molecular Formula C3H5BrMg
Molecular Weight (g/mol) 145.28
MDL Number MFCD00209610
InChI Key HHNLJRLNDSEYOX-UHFFFAOYSA-M
Synonym 1-propenylmagnesium bromide, propenylmagnesium bromide, 1-propenyl magnesium bromide, magnesium, bromo-1z-1-propenyl, e-prop-1-en-1-ylmagnesium bromide, propenyl magnesium bromide, z-propenylmagnesium bromide, prop-1-enylmagnesium bromide, e-propenylmagnesium bromide, prop-1-en-1-ylmagnesium bromide, grignard reagent
PubChem CID 5112801
IUPAC Name magnesium;prop-1-ene;bromide
SMILES C\C=C\[Mg]Br

Specifications

CAS Min % 6.88, 91.59
CAS Max % 8.41, 93.12
Color Brown to Yellow
Density 0.9500g/mL
Boiling Point 65.0°C
Flash Point −20°C
Packaging AcroSeal™ Glass Bottle
Quantity 800 mL
Specific Gravity 0.95
Solubility Information Solubility in water: reacts.
Formula Weight 145.28
Physical Form Solution
Chemical Name or Material 1-Propenylmagnesium bromide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Reacts violently w
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Handle under inert gas.
Protect from moisture

RUO – Research Use Only