accessibility menu, dialog, popup

UserName

CH3MgBr(E)

CAS RN 14092-04-7

IUPAC Name: bromo[(1E)-prop-1-en-1-yl]magnesium
Synonyms: bromo((1E)-prop-1-en-1-yl)magnesium
Molecular Weight (g/mol): 145.28
Molecular Formula: C3H5BrMg
InChi Key: HHNLJRLNDSEYOX-UHFFFAOYSA-M
SMILES: C\C=C\[Mg]Br
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)