missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(Phenylsulfonyl)pyrrole 98.0+%, TCI America™
$36.57 - $57.75
Chemical Identifiers
| CAS | 16851-82-4 |
|---|---|
| Molecular Formula | C10H9NO2S |
| Molecular Weight (g/mol) | 207.25 |
| MDL Number | MFCD00067739 |
| InChI Key | PPPXRIUHKCOOMU-UHFFFAOYSA-N |
| PubChem CID | 140146 |
| IUPAC Name | 1-(benzenesulfonyl)-1H-pyrrole |
| SMILES | O=S(=O)(N1C=CC=C1)C1=CC=CC=C1 |
Chemical Identifiers
| 16851-82-4 | |
| 207.25 | |
| PPPXRIUHKCOOMU-UHFFFAOYSA-N | |
| 1-(benzenesulfonyl)-1H-pyrrole |
| C10H9NO2S | |
| MFCD00067739 | |
| 140146 | |
| O=S(=O)(N1C=CC=C1)C1=CC=CC=C1 |
Specifications
| 16851-82-4 | |
| White | |
| MFCD00067739 | |
| 1-phenylsulfonyl pyrrole, 1-phenylsulfonyl-1h-pyrrole, n-benzenesulphonylpyrrole, 1-benzenesulfonyl pyrrole, n-benzenesulfonylpyrrole, 1-benzenesulfonyl-1h-pyrrole, n-phenylsulfonylpyrrole, phenylsulfonyl pyrrole, 1h-pyrrole,1-phenylsulfonyl, 1h-pyrrole, 1-phenylsulfonyl | |
| O=S(=O)(N1C=CC=C1)C1=CC=CC=C1 | |
| 207.25 | |
| 207.25 | |
| Crystalline Powder |
| 89°C | |
| C10H9NO2S | |
| 1 g | |
| PPPXRIUHKCOOMU-UHFFFAOYSA-N | |
| 1-(benzenesulfonyl)-1H-pyrrole | |
| 140146 | |
| ≥98.0% (GC) | |
| 1-(Phenylsulfonyl)pyrrole |
Safety and Handling
TSCA : No