missing translation for 'onlineSavingsMsg'
Learn More

1-(Phenylsulfonyl)indole, 98%

Catalog No. AAL1756603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1756603 1 g
AAL1756606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1756603 Supplier Thermo Scientific Chemicals Supplier No. L1756603

CAS: 40899-71-6 | C14H11NO2S | 257.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 40899-71-6
Molecular Formula C14H11NO2S
Molecular Weight (g/mol) 257.31
MDL Number MFCD00134318
InChI Key VDWLCYCWLIKWBV-UHFFFAOYSA-N
Synonym 1-phenylsulfonyl-1h-indole, 1-phenylsulfonyl indole, 1-benzenesulfonyl indole, 1-benzenesulfonyl-1h-indole, n-phenylsulfonylindole, 1-phenylsulphonyl indole, 1-phenylsulfonyl-indole, chembl82224
PubChem CID 315017
IUPAC Name 1-(benzenesulfonyl)indole
SMILES O=S(=O)(N1C=CC2=CC=CC=C12)C1=CC=CC=C1

Specifications

Melting Point 77°C to 79°C
Boiling Point 192°C (3 mmHg)
Quantity 1 g
Beilstein 1286712
Formula Weight 257.31
Percent Purity 98%
Chemical Name or Material 1-(Phenylsulfonyl)indole
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only