CAS RN 40899-71-6
IUPAC Name:
1-(benzenesulfonyl)-1H-indole
Synonyms:
1-(benzenesulfonyl)indole
Molecular Weight (g/mol):
257.31
Molecular Formula:
C14H11NO2S
InChi Key:
VDWLCYCWLIKWBV-UHFFFAOYSA-N
SMILES:
O=S(=O)(N1C=CC2=CC=CC=C12)C1=CC=CC=C1
1
–
1
of
1
results
1
Promotions Available
1-(Phenylsulfonyl)indole, 98%
1-(Phenylsulfonyl)indole, 98%
CAS: 40899-71-6 Molecular Formula: C14H11NO2S Molecular Weight (g/mol): 257.31 MDL Number: MFCD00134318 InChI Key: VDWLCYCWLIKWBV-UHFFFAOYSA-N Synonym: 1-phenylsulfonyl-1h-indole,1-phenylsulfonyl indole,1-benzenesulfonyl indole,1-benzenesulfonyl-1h-indole,n-phenylsulfonylindole,1-phenylsulphonyl indole,1-phenylsulfonyl-indole,chembl82224 PubChem CID: 315017 IUPAC Name: 1-(benzenesulfonyl)indole SMILES: O=S(=O)(N1C=CC2=CC=CC=C12)C1=CC=CC=C1