missing translation for 'onlineSavingsMsg'
Learn More
Learn More
[1-(Phenylsulfonyl)-1H-indol-3-yl]methanol, ≥97%, Thermo Scientific™
$193.86 - $193.86
Chemical Identifiers
| CAS | 89241-33-8 |
|---|---|
| Molecular Formula | C15H13NO3S |
| Molecular Weight (g/mol) | 287.333 |
| MDL Number | MFCD02682024 |
| InChI Key | ZMLXSFMIPYDHIN-UHFFFAOYSA-N |
| Synonym | 1-phenylsulfonyl-1h-indol-3-yl methanol, 1-benzenesulfonyl indol-3-yl methanol, 1h-indole-3-methanol,1-phenylsulfonyl, 1-benzenesulfonyl-1h-indol-3-yl methanol, acmc-20ljqe, 1-benzenesulfonyl-1h-indol-3-ylmethanol, 3-hydroxymethyl-n-phenylsulfonyl indole, 1-phenylsulfonyl-1h-indole-3-methanol |
| PubChem CID | 2776213 |
| IUPAC Name | [1-(benzenesulfonyl)indol-3-yl]methanol |
| SMILES | C1=CC=C(C=C1)S(=O)(=O)N2C=C(C3=CC=CC=C32)CO |
Chemical Identifiers
| 89241-33-8 | |
| 287.333 | |
| ZMLXSFMIPYDHIN-UHFFFAOYSA-N | |
| 2776213 | |
| C1=CC=C(C=C1)S(=O)(=O)N2C=C(C3=CC=CC=C32)CO |
| C15H13NO3S | |
| MFCD02682024 | |
| 1-phenylsulfonyl-1h-indol-3-yl methanol, 1-benzenesulfonyl indol-3-yl methanol, 1h-indole-3-methanol,1-phenylsulfonyl, 1-benzenesulfonyl-1h-indol-3-yl methanol, acmc-20ljqe, 1-benzenesulfonyl-1h-indol-3-ylmethanol, 3-hydroxymethyl-n-phenylsulfonyl indole, 1-phenylsulfonyl-1h-indole-3-methanol | |
| [1-(benzenesulfonyl)indol-3-yl]methanol |
Specifications
| 89241-33-8 | |
| 100.0 | |
| Amber Glass Bottle | |
| MFCD02682024 | |
| 1-phenylsulfonyl-1h-indol-3-yl methanol, 1-benzenesulfonyl indol-3-yl methanol, 1h-indole-3-methanol,1-phenylsulfonyl, 1-benzenesulfonyl-1h-indol-3-yl methanol, acmc-20ljqe, 1-benzenesulfonyl-1h-indol-3-ylmethanol, 3-hydroxymethyl-n-phenylsulfonyl indole, 1-phenylsulfonyl-1h-indole-3-methanol | |
| C1=CC=C(C=C1)S(=O)(=O)N2C=C(C3=CC=CC=C32)CO | |
| 287.333 | |
| 287.34 | |
| [1-(Phenylsulfonyl)-1H-indol-3-yl]methanol |
| 96.5 | |
| 97% | |
| C15H13NO3S | |
| 1 g | |
| ZMLXSFMIPYDHIN-UHFFFAOYSA-N | |
| [1-(benzenesulfonyl)indol-3-yl]methanol | |
| 2776213 | |
| 97% |