CAS RN 89241-33-8
IUPAC Name:
[1-(benzenesulfonyl)-1H-indol-3-yl]methanol
Synonyms:
[1-(benzenesulfonyl)indol-3-yl]methanol
Molecular Weight (g/mol):
287.33
Molecular Formula:
C15H13NO3S
InChi Key:
ZMLXSFMIPYDHIN-UHFFFAOYSA-N
SMILES:
OCC1=CN(C2=CC=CC=C12)S(=O)(=O)C1=CC=CC=C1
1
–
2
of
2
results
1
[1-(Phenylsulfonyl)-1H-indol-3-yl]methanol, ≥97%, Thermo Scientific™
CAS: 89241-33-8 Molecular Formula: C15H13NO3S Molecular Weight (g/mol): 287.333 MDL Number: MFCD02682024 InChI Key: ZMLXSFMIPYDHIN-UHFFFAOYSA-N Synonym: 1-phenylsulfonyl-1h-indol-3-yl methanol,1-benzenesulfonyl indol-3-yl methanol,1h-indole-3-methanol,1-phenylsulfonyl,1-benzenesulfonyl-1h-indol-3-yl methanol,acmc-20ljqe,1-benzenesulfonyl-1h-indol-3-ylmethanol,3-hydroxymethyl-n-phenylsulfonyl indole,1-phenylsulfonyl-1h-indole-3-methanol PubChem CID: 2776213 IUPAC Name: [1-(benzenesulfonyl)indol-3-yl]methanol SMILES: C1=CC=C(C=C1)S(=O)(=O)N2C=C(C3=CC=CC=C32)CO