missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Phenyl-3-methyl-5-pyrazolone, Spectrum™ Chemical
P2352, 89-25-8, C10H10N2O
$264.63 - $264.63
Chemical Identifiers
| CAS | 89-25-8 |
|---|---|
| Molecular Formula | C10H10N2O |
| Molecular Weight (g/mol) | 174.20 |
| InChI Key | QELUYTUMUWHWMC-UHFFFAOYSA-N |
| IUPAC Name | 3-methyl-1-phenyl-4,5-dihydro-1H-pyrazol-5-one |
| SMILES | CC1=NN(C(=O)C1)C1=CC=CC=C1 |
Description
Spectrum™ Chemical 1-Phenyl-3-methyl-5-pyrazoloneChemical Identifiers
| 89-25-8 | |
| 174.20 | |
| 3-methyl-1-phenyl-4,5-dihydro-1H-pyrazol-5-one |
| C10H10N2O | |
| QELUYTUMUWHWMC-UHFFFAOYSA-N | |
| CC1=NN(C(=O)C1)C1=CC=CC=C1 |
Specifications
| 89-25-8 | |
| Poly Bottle | |
| 500 g | |
| CC1=NN(C(=O)C1)C1=CC=CC=C1 | |
| 174.20 |
| 100% | |
| C10H10N2O | |
| QELUYTUMUWHWMC-UHFFFAOYSA-N | |
| 3-methyl-1-phenyl-4,5-dihydro-1H-pyrazol-5-one | |
| Reagent |