accessibility menu, dialog, popup

UserName

CH3NNO

CAS RN 89-25-8

IUPAC Name: 3-methyl-1-phenyl-4,5-dihydro-1H-pyrazol-5-one
Synonyms: edaravone 5-methyl-2-phenylpyrazol-3-one norphenazone 1-phenyl-3-methylpyrazolone 1-phenyl-3-methylpyrazolone-5 norantipyrine methylphenylpyrazolone 3-methyl-1-phenyl-5-pyrazolone ccris 512 3-methyl-1-phenylpyrazol-5-one
Molecular Weight (g/mol): 174.2
Molecular Formula: C10H10N2O
InChi Key: QELUYTUMUWHWMC-UHFFFAOYSA-N
SMILES: CC1=NN(C(=O)C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (1)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)
Boiling Point 
  • (3)
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
  • (3)
Physical Form 
  • (5)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (1)
  • (1)
  • (2)
  • (6)
  • (3)
  • (1)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (5)
  • (1)
  • (1)