missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Phenyl-3-methyl-5-pyrazolone, Spectrum™ Chemical
Description
Spectrum™ Chemical 1-Phenyl-3-methyl-5-pyrazolone
Specifications
Specifications
| CAS Max % | 100% |
| Packaging | Poly Bottle |
| Quantity | 500 g |
| Grade | Ungraded |
| Physical Form | Crystalline Powder or Lumps |
| Chemical Name or Material | 1-Phenyl-3-methyl-5-pyrazolone |