missing translation for 'onlineSavingsMsg'
Learn More

1-(p-Tolyl)piperazine 98.0+%, TCI America™

Catalog No. T29565G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T29565G 5 g
T295625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T29565G Supplier TCI America Supplier No. T29565G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 39593-08-3
Molecular Formula C11H16N2
Molecular Weight (g/mol) 176.263
MDL Number MFCD00040737
InChI Key ONEYFZXGNFNRJH-UHFFFAOYSA-N
Synonym 1-4-methylphenyl piperazine, 1-p-tolyl piperazine, 1-p-tolyl-piperazine, 1-4-methylphenyl-piperazine, 1-4-methyl phenyl piperazine, 1-p-tolylpiperazine, acmc-1adtx, 4-methylphenyl piperazine, 4-4-methylphenyl piperazine, 1-4-methyl-phenyl-piperazine
PubChem CID 83113
IUPAC Name 1-(4-methylphenyl)piperazine
SMILES CC1=CC=C(C=C1)N2CCNCC2

Specifications

Melting Point 36°C
Color White-Yellow
Boiling Point 112°C
Quantity 5 g
UN Number 3263
Formula Weight 176.26
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 1-(p-Tolyl)piperazine