missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(p-Toluenesulfonyl)imidazole 98.0+%, TCI America™
$68.97 - $244.19
Chemical Identifiers
| CAS | 2232-08-8 |
|---|---|
| Molecular Formula | C10H10N2O2S |
| Molecular Weight (g/mol) | 222.26 |
| MDL Number | MFCD00005285 |
| InChI Key | YJYMYJRAQYREBT-UHFFFAOYSA-N |
| Synonym | 1-p-toluenesulfonyl imidazole, 1-tosyl-1h-imidazole, n-tosylimidazole, 1-4-methylphenyl sulfonyl-1h-imidazole, 1-tosylimidazole, 1-p-toluenesulphonyl imidazole, 1-p-tolylsulfonyl imidazole, 1h-imidazole, 1-4-methylphenyl sulfonyl, 1-toluene-p-sulphonyl imidazole, 1-4-methylbenzenesulfonyl imidazole |
| PubChem CID | 75219 |
| IUPAC Name | 1-(4-methylbenzenesulfonyl)-1H-imidazole |
| SMILES | CC1=CC=C(C=C1)S(=O)(=O)N1C=CN=C1 |
Chemical Identifiers
| 2232-08-8 | |
| 222.26 | |
| YJYMYJRAQYREBT-UHFFFAOYSA-N | |
| 75219 | |
| CC1=CC=C(C=C1)S(=O)(=O)N1C=CN=C1 |
| C10H10N2O2S | |
| MFCD00005285 | |
| 1-p-toluenesulfonyl imidazole, 1-tosyl-1h-imidazole, n-tosylimidazole, 1-4-methylphenyl sulfonyl-1h-imidazole, 1-tosylimidazole, 1-p-toluenesulphonyl imidazole, 1-p-tolylsulfonyl imidazole, 1h-imidazole, 1-4-methylphenyl sulfonyl, 1-toluene-p-sulphonyl imidazole, 1-4-methylbenzenesulfonyl imidazole | |
| 1-(4-methylbenzenesulfonyl)-1H-imidazole |
Specifications
| 2232-08-8 | |
| White-Yellow | |
| MFCD00005285 | |
| 1-p-toluenesulfonyl imidazole, 1-tosyl-1h-imidazole, n-tosylimidazole, 1-4-methylphenyl sulfonyl-1h-imidazole, 1-tosylimidazole, 1-p-toluenesulphonyl imidazole, 1-p-tolylsulfonyl imidazole, 1h-imidazole, 1-4-methylphenyl sulfonyl, 1-toluene-p-sulphonyl imidazole, 1-4-methylbenzenesulfonyl imidazole | |
| CC1=CC=C(C=C1)S(=O)(=O)N1C=CN=C1 | |
| 222.26 | |
| 222.26 | |
| Crystalline Powder |
| 77°C | |
| C10H10N2O2S | |
| 5 g | |
| YJYMYJRAQYREBT-UHFFFAOYSA-N | |
| 1-(4-methylbenzenesulfonyl)-1H-imidazole | |
| 75219 | |
| ≥98.0% (HPLC,N) | |
| 1-(p-Toluenesulfonyl)imidazole |
Safety and Handling
TSCA : No