accessibility menu, dialog, popup

UserName

CH3SOONN

CAS RN 2232-08-8

IUPAC Name: 1-(4-methylbenzenesulfonyl)-1H-imidazole
Synonyms: 1-(4-methylbenzenesulfonyl)imidazole
Molecular Weight (g/mol): 222.26
Molecular Formula: C10H10N2O2S
InChi Key: YJYMYJRAQYREBT-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)N1C=CN=C1
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)