missing translation for 'onlineSavingsMsg'
Learn More

1-(p-Toluenesulfonyl)imidazole, 98+%

Catalog No. p-7035957
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5419106 5 g
AAH5419114 25 g
AAH5419122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH5419106 Supplier Thermo Scientific Chemicals Supplier No. H5419106

CAS: 2232-08-8 | C10H10N2O2S | 222.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2232-08-8
Molecular Formula C10H10N2O2S
Molecular Weight (g/mol) 222.26
MDL Number MFCD00005285
InChI Key YJYMYJRAQYREBT-UHFFFAOYSA-N
Synonym 1-p-toluenesulfonyl imidazole, 1-tosyl-1h-imidazole, n-tosylimidazole, 1-4-methylphenyl sulfonyl-1h-imidazole, 1-tosylimidazole, 1-p-toluenesulphonyl imidazole, 1-p-tolylsulfonyl imidazole, 1h-imidazole, 1-4-methylphenyl sulfonyl, 1-toluene-p-sulphonyl imidazole, 1-4-methylbenzenesulfonyl imidazole
PubChem CID 75219
IUPAC Name 1-(4-methylphenyl)sulfonylimidazole
SMILES CC1=CC=C(C=C1)S(=O)(=O)N1C=CN=C1

Specifications

Melting Point 76°C to 78°C
Quantity 5 g
Beilstein 612451
Sensitivity Moisture sensitive
Formula Weight 222.26
Percent Purity ≥98%
Chemical Name or Material 1-(p-Toluenesulfonyl)imidazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 218-771-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only