missing translation for 'onlineSavingsMsg'
Learn More

1-Octyl isothiocyanate, 97%

Catalog No. p-7021954
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1450106 5 g
AAA1450114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1450106 Supplier Thermo Scientific Chemicals Supplier No. A1450106
Only null left

CAS: 4430-45-9 | C9H17NS | 171.30 g/mol

1-Octyl isothiocyanate can be used to produce 1-(6-methyl-quinolin-2-yl)-3-octyl-thiourea. It is used as intermediates in pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Octyl isothiocyanate can be used to produce 1-(6-methyl-quinolin-2-yl)-3-octyl-thiourea. It is used as intermediates in pharmaceutical industry.

Solubility
It hydrolyzes in water.

Notes
It is moisture sensitive. Incompatible with oxidizing agents, alcohols and amines.

Chemical Identifiers

CAS 4430-45-9
Molecular Formula C9H17NS
Molecular Weight (g/mol) 171.30
MDL Number MFCD00041139
InChI Key YEZHGQZHWKJPCM-UHFFFAOYSA-N
Synonym octyl isothiocyanate, n-octyl isothiocyanate, octylisothiocyanate, 1-octyl isothiocyanate, octanisothiocyanate, 1-isothiocyanatooctan, octane, 1-isothiocyanato, isothiocyanic acid, octyl ester, n-octylisothiocyanate, acmc-1ajcy
PubChem CID 78161
IUPAC Name 1-isothiocyanatooctane
SMILES CCCCCCCCN=C=S

Specifications

Density 0.88
Boiling Point 124°C to 126°C (7 mmHg)
Flash Point 69°C (156°F)
Odor Pungent
Refractive Index 1.49
Quantity 5 g
UN Number UN2922
Beilstein 1760783
Sensitivity Moisture sensitive
Solubility Information It hydrolyzes in water.
Formula Weight 171.31
Percent Purity 97%
Chemical Name or Material 1-Octyl isothiocyanate
Show More Show Less
Hazard Category H227-H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H314-H318-H227-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 224-625-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only