accessibility menu, dialog, popup

UserName

CAS RN 4430-45-9

IUPAC Name: 1-isothiocyanatooctane
Synonyms: 1-isothiocyanatooctane
Molecular Weight (g/mol): 171.3
Molecular Formula: C9H17NS
InChi Key: YEZHGQZHWKJPCM-UHFFFAOYSA-N
SMILES: CCCCCCCCN=C=S
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)