missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(o-Tolyl)biguanide 97.0+%, TCI America™
Supplier: TCI America T0314500G
Specifications
| 1-(o-Tolyl)biguanide | |
| 147°C | |
| C9H13N5 | |
| 500 g | |
| CC1=CC=CC=C1N=C(N)N=C(N)N | |
| 191.238 | |
| 191.24 | |
| Crystalline Powder |
| 93-69-6 | |
| White | |
| MFCD00019731 | |
| SQZCAOHYQSOZCE-UHFFFAOYSA-N | |
| 1-(diaminomethylidene)-2-(2-methylphenyl)guanidine | |
| 7155 | |
| ≥97.0% (T) |
Chemical Identifiers
| 93-69-6 | |
| 191.238 | |
| SQZCAOHYQSOZCE-UHFFFAOYSA-N | |
| 1-(diaminomethylidene)-2-(2-methylphenyl)guanidine |
| C9H13N5 | |
| MFCD00019731 | |
| 7155 | |
| CC1=CC=CC=C1N=C(N)N=C(N)N |
Safety and Handling
EINECSNumber : (3)-2191&(3)-3215&(3)-0260
RTECSNumber : DU2800000
TSCA : Yes