missing translation for 'onlineSavingsMsg'
Learn More

1-(o-Tolyl)biguanide 97.0+%, TCI America™

Catalog No. p-7136214
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T031425G 25 g
T0314500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T031425G Supplier TCI America Supplier No. T031425G
Only null left

Chemical Identifiers

CAS 93-69-6
Molecular Formula C9H13N5
Molecular Weight (g/mol) 191.238
MDL Number MFCD00019731
InChI Key SQZCAOHYQSOZCE-UHFFFAOYSA-N
PubChem CID 7155
IUPAC Name 1-(diaminomethylidene)-2-(2-methylphenyl)guanidine
SMILES CC1=CC=CC=C1N=C(N)N=C(N)N

Specifications

Melting Point 147°C
Color White
Quantity 25 g
Formula Weight 191.24
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(o-Tolyl)biguanide
EINECSNumber (3)-2191&(3)-3215&(3)-0260
RTECSNumber DU2800000
TSCA Yes