accessibility menu, dialog, popup

UserName

CAS RN 93-69-6

IUPAC Name: N-(diaminomethylidene)-N''-(2-methylphenyl)guanidine
Synonyms: O-tolyl biguanide
Molecular Weight (g/mol): 191.24
Molecular Formula: C9H13N5
InChi Key: SQZCAOHYQSOZCE-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1N=C(N)N=C(N)N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results