missing translation for 'onlineSavingsMsg'
Learn More

1-O-n-Octyl-beta-D-glucopyranoside, 98%

Catalog No. AC230340010
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC230340010 1 g Glass bottle
AC230340050 5 g Glass bottle
AC230340250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC230340010 Supplier Thermo Scientific Chemicals Supplier No. 230340010

CAS: 29836-26-8 | C14H28O6 | 292.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 29836-26-8
Molecular Formula C14H28O6
Molecular Weight (g/mol) 292.37
MDL Number MFCD00063288
InChI Key HEGSGKPQLMEBJL-RKQHYHRCSA-N
Synonym octyl-beta-d-glucopyranoside, b-octylglucoside, octyl beta-d-glucopyranoside, octyl beta-d-glucoside, octyl glucoside, n-octyl-beta-d-glucoside, octyl b-d-glucopyranoside, 2r,3s,4s,5r,6r-2-hydroxymethyl-6-octyloxy tetrahydro-2h-pyran-3,4,5-triol, unii-v109wut6rl, n-octyl glucoside
PubChem CID 62852
IUPAC Name (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-octoxyoxane-3,4,5-triol
SMILES CCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 105°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (ELSD) (HPLC)
Packaging Glass bottle
Quantity 1 g
Merck Index 15, 6857
Specific Rotation Condition − 32.00 (20.00°C c=5,water)
Specific Rotation -32
Solubility Information (10% in water) Clear to slightly hazy colorless to light yellow
Formula Weight 292.36
Percent Purity 98%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 1-O-n-Octyl-β-D-glucopyranoside
Show More Show Less
EINECSNumber 249-887-8

RUO – Research Use Only