missing translation for 'onlineSavingsMsg'
Learn More

1-Nitropropane 98.0+%, TCI America™

Catalog No. N024825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N024825G 25 g
N0248500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N024825G Supplier TCI America Supplier No. N024825G

Chemical Identifiers

CAS 108-03-2
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.094
MDL Number MFCD00007407
InChI Key JSZOAYXJRCEYSX-UHFFFAOYSA-N
Synonym propane, 1-nitro, nitropropane, propane, nitro, ccris 1329, dsstox_cid_980, dsstox_rid_75902, dsstox_gsid_20980, nitro-propane, n-nitropropane, 3-nitropropane
PubChem CID 7903
ChEBI CHEBI:76261
IUPAC Name 1-nitropropane
SMILES CCC[N+](=O)[O-]

Specifications

Color Yellow
Boiling Point 131°C
Quantity 25 g
UN Number 2608
Formula Weight 89.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Nitropropane
EINECSNumber (2)-3738
RTECSNumber TZ5075000
TSCA Yes