missing translation for 'onlineSavingsMsg'
Learn More

1-Nitro-3-(trifluoromethoxy)benzene 98.0+%, TCI America™

Catalog No. p-7134010
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
N08845G 5 g
N088425G 25 g
2 options

Catalog No. N08845G

Supplier: TCI America N08845G

Only null left

Chemical Identifiers

CAS 2995-45-1
Molecular Formula C7H4F3NO3
Molecular Weight (g/mol) 207.11
MDL Number MFCD00041011
InChI Key QBWJNDOQIAARBT-UHFFFAOYSA-N
PubChem CID 2777297
IUPAC Name 1-nitro-3-(trifluoromethoxy)benzene
SMILES [O-][N+](=O)C1=CC(OC(F)(F)F)=CC=C1

Specifications

Color Yellow
Boiling Point 96°C
Quantity 5 g
Formula Weight 207.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Nitro-3-(trifluoromethoxy)benzene