accessibility menu, dialog, popup

UserName

CAS RN 2995-45-1

OFFFNOO

CAS RN 2995-45-1

IUPAC Name: 1-nitro-3-(trifluoromethoxy)benzene
Synonyms: 1-nitro-3-(trifluoromethoxy)benzene
Molecular Weight (g/mol): 207.11
Molecular Formula: C7H4F3NO3
InChi Key: QBWJNDOQIAARBT-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(OC(F)(F)F)=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results