missing translation for 'onlineSavingsMsg'
Learn More

1-Nitro-2-naphthol 98.0+%, TCI America™

Catalog No. N03701G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N03701G 1 g
N03705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N03701G Supplier TCI America Supplier No. N03701G

Chemical Identifiers

CAS 550-60-7
Molecular Formula C10H7NO3
Molecular Weight (g/mol) 189.17
MDL Number MFCD00021461
InChI Key SSHIVHKMGVBXTJ-UHFFFAOYSA-N
Synonym 2-Hydroxy-1-nitronaphthalene
PubChem CID 11075
IUPAC Name 1-nitronaphthalen-2-ol
SMILES C1=CC=C2C(=C1)C=CC(=C2[N+](=O)[O-])O

Specifications

Melting Point 103°C
Quantity 1 g
Formula Weight 189.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Nitro-2-naphthol
RTECSNumber QL4500000
TSCA No