accessibility menu, dialog, popup

UserName

HONOO

CAS RN 550-60-7

IUPAC Name: 1-nitronaphthalen-2-ol
Synonyms: 1-nitro-2-naphthol
Molecular Weight (g/mol): 189.17
Molecular Formula: C10H7NO3
InChi Key: SSHIVHKMGVBXTJ-UHFFFAOYSA-N
SMILES: OC1=CC=C2C=CC=CC2=C1[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results