missing translation for 'onlineSavingsMsg'
Learn More

1-Nitro-2-(n-octyloxy)benzene, 98%

Catalog No. AAA1732006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1732006 5 g
AAA1732014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1732006 Supplier Thermo Scientific Chemicals Supplier No. A1732006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 37682-29-4 | C14H21NO3 | 251.326 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 37682-29-4
Molecular Formula C14H21NO3
Molecular Weight (g/mol) 251.326
MDL Number MFCD00014693
InChI Key CXVOIIMJZFREMM-UHFFFAOYSA-N
Synonym 2-nitrophenyl octyl ether, 1-nitro-2-octyloxy benzene, octyl o-nitrophenyl ether, 2-nitrophenyl n-octyl ether, 2-octyloxy nitrobenzene, benzene, 1-nitro-2-octyloxy, 1-2-nitrophenoxy octane, 1-nitro-2-octyloxybenzene, 2-n-octyloxy nitrobenzene, npoe
PubChem CID 169952
IUPAC Name 1-nitro-2-octoxybenzene
SMILES CCCCCCCCOC1=CC=CC=C1[N+](=O)[O-]

Specifications

Density 1.04
Boiling Point 163°C to 165°C (2 mmHg)
Flash Point 113°C (235°F)
Refractive Index 1.511
Quantity 5 g
Beilstein 2529905
Formula Weight 251.32
Percent Purity 98%
Chemical Name or Material 1-Nitro-2-(n-octyloxy)benzene
EINECSNumber 253-623-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only