missing translation for 'onlineSavingsMsg'
Learn More

(1-Naphthylmethyl)triphenylphosphonium Chloride 98.0+%, TCI America™

Catalog No. n07005g
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N07005G 5 g
N070025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N07005G Supplier TCI America Supplier No. N07005G

Chemical Identifiers

CAS 23277-00-1
Molecular Formula C29H24ClP
Molecular Weight (g/mol) 438.93
MDL Number MFCD00031658
InChI Key MOYSMPXSEXYEJV-UHFFFAOYSA-M
Synonym 1-naphthylmethyl triphenylphosphonium chloride, naphthalen-1-ylmethyl triphenylphosphonium chloride, 1-naphthylmethyl triphenylphosphoniumchloride, naphthalen-1-ylmethyl triphenyl phosphanium chloride, 1-naphthylmethyltriphenylphosphonium chloride, acmc-209g2f, 1-napthylmethyl triphenylphosphonium chloride, triphenyl 1-naphthylmethyl phosphonium chloride, 1-naphthylmethyl-triphenylphosphonium chloride, 1-naphthalenylmethyl triphenyl phosphonium chloride
PubChem CID 2734113
IUPAC Name [(naphthalen-1-yl)methyl]triphenylphosphanium chloride
SMILES [Cl-].C(C1=CC=CC2=CC=CC=C12)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 438.93
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (1-Naphthylmethyl)triphenylphosphonium Chloride