accessibility menu, dialog, popup

UserName

CAS RN 23277-00-1

IUPAC Name: [(naphthalen-1-yl)methyl]triphenylphosphanium chloride
Synonyms: (naphthalen-1-ylmethyl)triphenylphosphanium chloride
Molecular Weight (g/mol): 438.93
Molecular Formula: C29H24ClP
InChi Key: MOYSMPXSEXYEJV-UHFFFAOYSA-M
SMILES: [Cl-].C(C1=CC=CC2=CC=CC=C12)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results