missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthoic acid, 98%

Catalog No. p-7080655
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1027314 25 g
AAA1027322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1027314 Supplier Thermo Scientific Chemicals Supplier No. A1027314

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 86-55-5 | C11H8O2 | 172.183 g/mol

1-Naphthoic acid is used as an intermediate for the synthesis of pharmaceuticals, photochemicals, plant growth hormones, dyes and other organic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Naphthoic acid is used as an intermediate for the synthesis of pharmaceuticals, photochemicals, plant growth hormones, dyes and other organic compounds.

Solubility
Freely soluble in hot alcohol and ether. Slightly soluble in hot water.

Notes
Store away from oxidizing agents. Store in a cool, dry condition in well sealed containers.

Chemical Identifiers

CAS 86-55-5
Molecular Formula C11H8O2
Molecular Weight (g/mol) 172.183
MDL Number MFCD00004007
InChI Key LNETULKMXZVUST-UHFFFAOYSA-N
Synonym 1-naphthoic acid, 1-naphthalenecarboxylic acid, 1-carboxynaphthalene, naphthoic acid, alpha-naphthoic acid, naphthalenecarboxylic acid, carboxynaphthalene, alpha-naphthylcarboxylic acid, naphthalene-alpha-carboxylic acid, unii-2niv4o66bh
PubChem CID 6847
ChEBI CHEBI:36466
IUPAC Name naphthalene-1-carboxylic acid
SMILES C1=CC=C2C(=C1)C=CC=C2C(=O)O

Specifications

Melting Point 160°C to 162°C
Density 1.398
Boiling Point >300°C
Flash Point 195°C (383°F)
Quantity 25 g
Beilstein 1908896
Merck Index 14,6381
Solubility Information Freely soluble in hot alcohol and ether. Slightly soluble in hot water.
Formula Weight 172.18
Percent Purity 98%
Chemical Name or Material 1-Naphthoic acid
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 201-681-9
RTECSNumber QL0960000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only