missing translation for 'onlineSavingsMsg'
Learn More

1-Methylnaphthalene 96.0+%, TCI America™

Supplier:  TCI America M0371100ML

 View more versions of this product

Catalog No. M0371100ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1-Methylnaphthalene
Yellow
C11H10
100 mL
alpha-methylnaphthalene, naphthalene, 1-methyl, methylnaphthalene, naphthalene, methyl, polymethylnaphthalene, 1-methyl-naphthalene, alpha-methyl naphthalenes, methyl naphthalene, 1-methyl naphthalene, fema number 3193
CC1=CC=CC2=CC=CC=C12
142.201
CHEBI:50717
≥96.0% (GC)
90-12-0
241°C
MFCD00004034
3082
QPUYECUOLPXSFR-UHFFFAOYSA-N
1-methylnaphthalene
7002
142.20
Liquid

Chemical Identifiers

90-12-0
142.201
QPUYECUOLPXSFR-UHFFFAOYSA-N
7002
1-methylnaphthalene
C11H10
MFCD00004034
alpha-methylnaphthalene, naphthalene, 1-methyl, methylnaphthalene, naphthalene, methyl, polymethylnaphthalene, 1-methyl-naphthalene, alpha-methyl naphthalenes, methyl naphthalene, 1-methyl naphthalene, fema number 3193
CHEBI:50717
CC1=CC=CC2=CC=CC=C12
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (4)-0080

RTECSNumber : QJ9630000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions