accessibility menu, dialog, popup

UserName

CH3

CAS RN 90-12-0

IUPAC Name: 1-methylnaphthalene
Synonyms: 1-methylnaphthalene fema number 3193 methyl naphthalenes (mixture of isomers)
Molecular Weight (g/mol): 142.2
Molecular Formula: C11H10
InChi Key: QPUYECUOLPXSFR-UHFFFAOYSA-N
SMILES: CC1=C2C=CC=CC2=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (6)
Boiling Point 
  • (2)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (3)
  • (5)
  • (3)
  • (6)
  • (2)
  • (3)
  • (3)
  • (3)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (6)
  • (1)