missing translation for 'onlineSavingsMsg'
Learn More

1-Methylnaphthalene 96.0+%, TCI America™

Catalog No. p-7132126
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M037125ML 25 mL
M0371100ML 100 mL
M0371500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M037125ML Supplier TCI America Supplier No. M037125ML
Only null left

Chemical Identifiers

CAS 90-12-0
Molecular Formula C11H10
Molecular Weight (g/mol) 142.201
MDL Number MFCD00004034
InChI Key QPUYECUOLPXSFR-UHFFFAOYSA-N
Synonym alpha-methylnaphthalene, naphthalene, 1-methyl, methylnaphthalene, naphthalene, methyl, polymethylnaphthalene, 1-methyl-naphthalene, alpha-methyl naphthalenes, methyl naphthalene, 1-methyl naphthalene, fema number 3193
PubChem CID 7002
ChEBI CHEBI:50717
IUPAC Name 1-methylnaphthalene
SMILES CC1=CC=CC2=CC=CC=C12

Specifications

Color Yellow
Boiling Point 241°C
Quantity 25 mL
UN Number 3082
Formula Weight 142.20
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Methylnaphthalene
EINECSNumber (4)-0080
RTECSNumber QJ9630000
TSCA Yes