missing translation for 'onlineSavingsMsg'
Learn More

1-Methylindole-3-carbonitrile, 96%

CAS: 24662-37-1 | C10H8N2 | 156.188 g/mol

Supplier:  Thermo Scientific Chemicals H3415703

 View more versions of this product

Catalog No. AAH3415703


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Methylindole-3-carbonitrile
55°C to 56°C
MFCD00466602
1-methyl-1h-indole-3-carbonitrile, 3-cyano-1-methylindole, 1h-indole-3-carbonitrile,1-methyl, 1h-indole-3-carbonitrile, 1-methyl, d06fsr, 3-cyano 1-methyl-1h-indole, fbaxzpmxgbnbpe-uhfffaoysa
CN1C=C(C2=CC=CC=C21)C#N
156.188
156.19
24662-37-1
C10H8N2
1 g
FBAXZPMXGBNBPE-UHFFFAOYSA-N
1-methylindole-3-carbonitrile
2307681
96%

Chemical Identifiers

24662-37-1
156.188
FBAXZPMXGBNBPE-UHFFFAOYSA-N
1-methylindole-3-carbonitrile
C10H8N2
MFCD00466602
2307681
CN1C=C(C2=CC=CC=C21)C#N
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only