missing translation for 'onlineSavingsMsg'
Learn More

1-Methylindole-3-carbonitrile, 96%

Catalog No. p-7032625
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
250 mg
5 g
Catalog No. Quantity
AAH34157MD 250 mg
AAH3415703 1 g
AAH3415706 5 g
3 options

Catalog No. AAH34157MD

Supplier: Thermo Scientific Chemicals H34157MD

Only null left

CAS: 24662-37-1 | C10H8N2 | 156.188 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 24662-37-1
Molecular Formula C10H8N2
Molecular Weight (g/mol) 156.188
MDL Number MFCD00466602
InChI Key FBAXZPMXGBNBPE-UHFFFAOYSA-N
PubChem CID 2307681
IUPAC Name 1-methylindole-3-carbonitrile
SMILES CN1C=C(C2=CC=CC=C21)C#N

Specifications

Melting Point 55°C to 56°C
Quantity 250 mg
Synonym 1-methyl-1h-indole-3-carbonitrile, 3-cyano-1-methylindole, 1h-indole-3-carbonitrile,1-methyl, 1h-indole-3-carbonitrile, 1-methyl, d06fsr, 3-cyano 1-methyl-1h-indole, fbaxzpmxgbnbpe-uhfffaoysa
Formula Weight 156.19
Percent Purity 96%
Chemical Name or Material 1-Methylindole-3-carbonitrile
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only