accessibility menu, dialog, popup

UserName

CAS RN 24662-37-1

IUPAC Name: 1-methyl-1H-indole-3-carbonitrile
Synonyms: 1-methylindole-3-carbonitrile
Molecular Weight (g/mol): 156.19
Molecular Formula: C10H8N2
InChi Key: FBAXZPMXGBNBPE-UHFFFAOYSA-N
SMILES: CN1C=C(C#N)C2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results