missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methyl-3-propylimidazolium Iodide 97.0+%, TCI America™
$61.61 - $180.97
Chemical Identifiers
| CAS | 119171-18-5 |
|---|---|
| Molecular Formula | C7H13IN2 |
| Molecular Weight (g/mol) | 252.10 |
| MDL Number | MFCD08276385 |
| InChI Key | IVCMUVGRRDWTDK-UHFFFAOYSA-M |
| Synonym | 1-methyl-3-propylimidazolium iodide, 1-propyl-3-methylimidazolium iodide, 1-methyl-3-n-propylimidazolium iodide, 1-methyl-3-propylimidazol-1-ium iodide, acmc-1c1fw, dsstox_cid_29063, dsstox_rid_83282, dsstox_gsid_49207, ksc491c6t, 1-methyl-3-propylimidazoliumiodide |
| PubChem CID | 12971008 |
| IUPAC Name | 3-methyl-1-propyl-1H-imidazol-3-ium iodide |
| SMILES | [I-].CCCN1C=C[N+](C)=C1 |
Chemical Identifiers
| 119171-18-5 | |
| 252.10 | |
| IVCMUVGRRDWTDK-UHFFFAOYSA-M | |
| 12971008 | |
| [I-].CCCN1C=C[N+](C)=C1 |
| C7H13IN2 | |
| MFCD08276385 | |
| 1-methyl-3-propylimidazolium iodide, 1-propyl-3-methylimidazolium iodide, 1-methyl-3-n-propylimidazolium iodide, 1-methyl-3-propylimidazol-1-ium iodide, acmc-1c1fw, dsstox_cid_29063, dsstox_rid_83282, dsstox_gsid_49207, ksc491c6t, 1-methyl-3-propylimidazoliumiodide | |
| 3-methyl-1-propyl-1H-imidazol-3-ium iodide |
Specifications
| 119171-18-5 | |
| MFCD08276385 | |
| 1-methyl-3-propylimidazolium iodide, 1-propyl-3-methylimidazolium iodide, 1-methyl-3-n-propylimidazolium iodide, 1-methyl-3-propylimidazol-1-ium iodide, acmc-1c1fw, dsstox_cid_29063, dsstox_rid_83282, dsstox_gsid_49207, ksc491c6t, 1-methyl-3-propylimidazoliumiodide | |
| [I-].CCCN1C=C[N+](C)=C1 | |
| 252.10 | |
| 252.10 | |
| Liquid |
| C7H13IN2 | |
| 5 g | |
| IVCMUVGRRDWTDK-UHFFFAOYSA-M | |
| 3-methyl-1-propyl-1H-imidazol-3-ium iodide | |
| 12971008 | |
| ≥97.0% (HPLC,T) | |
| 1-Methyl-3-propylimidazolium Iodide |
Safety and Handling
TSCA : No