accessibility menu, dialog, popup

UserName

CAS RN 119171-18-5

IUPAC Name: 3-methyl-1-propyl-1H-imidazol-3-ium iodide
Synonyms: 1-methyl-3-propylimidazol-1-ium iodide
Molecular Weight (g/mol): 252.1
Molecular Formula: C7H13IN2
InChi Key: IVCMUVGRRDWTDK-UHFFFAOYSA-M
SMILES: [I-].CCCN1C=C[N+](C)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)